Copper(I) cyanide

Supplier: BeanTown Chemical

213300-1KG 213300-250G
BT213300-1KGEA 99 USD
BT213300-1KG BT213300-250G
Copper(I) cyanide
Copper(I) cyanide

CAS: 544-92-3; EC No: 208-883-6; MDL No: MFCD00010975; RTECS: GL7150000 UN No: UN1587; Haz Class: 6.1; Packing Group: II Powder; Linear Formula: CuCN; MW: 89.56 Melting Point: 474° Density (g/mL): 2.92


  • RTECS: GL7150000
  • Powder

Formula: CCuN
Melting Pt: 474 °C
Density: 2.92 g/mL
MDL Number: MFCD00010975
CAS Number: 544-92-3
EINECS: 208-883-6
UN: 1587
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR