2-Chloro-2-methylbutane ≥95%
Supplier: BeanTown Chemical
213260-100G
213260-25G
213260-5G
BT213260-100GEA
235
USD
BT213260-100G
BT213260-25G
BT213260-5G
2-Chloro-2-methylbutane ≥95%
2-Chloro-2-methylbutane
CAS: 594-36-5; EC No: 209-836-2; MDL No: MFCD00000818
UN No: UN1107; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C5H11Cl; MW: 106.6
Melting Point: -73°; Boiling Point: 85-86°; Flash point: -10°C (14°F)
Density (g/mL): 0.866; Refractive Index: 1.405
- Refractive index: 1.405
- Liquid
Formula:
C₅H₁₁Cl MW: 106.6 g/mol Boiling Pt: 85…86 °C Melting Pt: –73 °C Density: 0.866 g/mL Flash Pt: –10 °C (14 °F) |
MDL Number:
MFCD00000818 CAS Number: 594-36-5 EINECS: 209-836-2 UN: 1107 ADR: 3,II |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...