Vanadyl(IV) acetylacetonate
Supplier: BeanTown Chemical
Synonyms:
Vanadium(IV) oxide acetylacetonate, Oxobis(2,4-pentanedionato-O,O')vanadium (IV), Vanadium(IV) bis(acetylacetonato)oxide, Vanadyl acetylacetonate, Di(2,4-pentandionato-O,O')vanadium(IV)oxid, Vanadium (IV) oxide bis(2,4-pentanedionate)
213245-10G
213245-250G
213245-50G
BT213245-10GEA
22.97
USD
BT213245-10G
BT213245-250G
BT213245-50G
Vanadyl(IV) acetylacetonate
Vanadyl(IV) acetylacetonate
CAS: 3153-26-2; EC No: 221-590-8; MDL No: MFCD00000032
UN No: UN3285; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: OV(C5H7O2)2; Molecular Formula: C10H14O5V ; MW: 265.16
Melting Point: 235° (decomposes); Boiling Point: 140°/13 mmHg
Density (g/mL): 1.4
Air Sensitive
- Air Sensitive
- Powder
Formula:
C₁₀H₁₄O₅V MW: 265.16 g/mol Boiling Pt: 140 °C/13 mmHg Melting Pt: 235 °C (dec.) Density: 1.4 g/mL |
MDL Number:
MFCD00000032 CAS Number: 3153-26-2 EINECS: 221-590-8 UN: 3285 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...