3-Methoxypropene ≥94%

Supplier: BeanTown Chemical

Synonyms: Allyl methyl ether

213195-10G 213195-50G
BT213195-10GEA 93.62 USD
BT213195-10G BT213195-50G
3-Methoxypropene ≥94%
3-Methoxypropene

CAS: 627-40-7; EC No: 210-997-6; MDL No: MFCD00008649 UN No: UN3271; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH2=CHCH2OCH3; Molecular Formula: C4H8O; MW: 72.11 Boiling Point: 42-43° Density (g/mL): 0.77; Refractive Index: 1.380


  • Refractive index: 1.380
  • Liquid

Formula: C₄H₈O
Boiling Pt: 42…43 °C
Density: 0.77 g/mL
MDL Number: MFCD00008649
CAS Number: 627-40-7
EINECS: 210-997-6
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Assay 94%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR