4-Bromo-m-anisidine ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Bromo-3-methoxyaniline, 5-Amino-2-bromoanisole, 4-Bromo-3-anisidine
213185-25G
213185-5G
BT213185-25GEA
484
USD
BT213185-25G
BT213185-5G
4-Bromo-m-anisidine ≥97%
4-Bromo-m-anisidine
CAS: 19056-40-7; MDL No: MFCD05664063
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C7H8BrNO; MW: 202.05
Melting Point: 96-100°
- Solid
Formula:
C₇H₈BrNO Melting Pt: 96…100 °C |
MDL Number:
MFCD05664063 CAS Number: 19056-40-7 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...