1,4-Cyclohexadiene ≥97% stabilised
Supplier: BeanTown Chemical
Synonyms:
Cyclohexa-1,4-diene
213140-100G
213140-25G
213140-5G
BT213140-100GEA
407.99
USD
BT213140-100G
BT213140-25G
BT213140-5G
1,4-Cyclohexadiene ≥97% stabilised
1,4-Cyclohexadiene
CAS: 628-41-1; EC No: 211-043-1; MDL No: MFCD00001535
UN No: UN3295; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C6H8; MW: 80.13
Boiling Point: 88-89°; Flash point: -7°C (19°F)
Density (g/mL): 0.847; Refractive Index: 1.472
- Refractive index: 1.472
- Liquid
Formula:
C₆H₈ MW: 80.13 g/mol Boiling Pt: 82 Melting Pt: -49.2 Density: 0.847 g/mL Flash Pt: –7 °C (19 °F) |
MDL Number:
MFCD00001535 CAS Number: 628-41-1 EINECS: 211-043-1 UN: 3295 ADR: 3 II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...