(Benzyl)triphenylphosphonium chloride ≥99%

Supplier: BeanTown Chemical

Synonyms: Benzyltriphenylphosphonium chloride

213135-100G 213135-25G 213135-500G
BT213135-100GEA 85.11 USD
BT213135-100G BT213135-25G BT213135-500G
(Benzyl)triphenylphosphonium chloride ≥99%
(Benzyl)triphenylphosphonium chloride

CAS: 1100-88-5; EC No: 214-154-3; MDL No: MFCD00011913; RTECS: TA2416500 UN No: UN3464; Haz Class: 6.1; Packing Group: II Powder; Molecular Formula: C25H22ClP; MW: 388.87 Melting Point: 300°; Flash point: 300°C (572°F) Hygroscopic


  • RTECS: TA2416500
  • Hygroscopic
  • Powder

Formula: C₂₅H₂₂ClP
MW: 388.88 g/mol
Melting Pt: >300 °C
Flash Pt: 300 °C (572 °F)
MDL Number: MFCD00011913
CAS Number: 1100-88-5
EINECS: 214-154-3
UN: 3464
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR