Dilithium oxide ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
213120-10G
213120-250G
213120-50G
BT213120-10GEA
75.72
USD
BT213120-10G
BT213120-250G
BT213120-50G
Dilithium oxide ≥99.5% (trace metals basis)
di-Lithium oxide
CAS: 12057-24-8; EC No: 235-019-5; MDL No: MFCD00016183
UN No: UN3262; Haz Class: 8; Packing Group: II
-100 Mesh Powder; Molecular Formula: Li2O ; MW: 29.88
Density (g/mL): 2.013
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- -100 Mesh Powder
Formula:
Li₂O Density: 2.013 g/mL |
MDL Number:
MFCD00016183 CAS Number: 12057-24-8 EINECS: 235-019-5 UN: 3262 ADR: 8,II |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...