Iron(III) fluoride trihydrate

Supplier: BeanTown Chemical

213100-250G 213100-50G
BT213100-250GEA 261 USD
BT213100-250G BT213100-50G
Iron(III) fluoride trihydrate
Iron(III) fluoride trihydrate

CAS: 15469-38-2; EC No: 232-002-4; MDL No: MFCD00149718 UN No: UN3260; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: FeF3·3H2O; MW: 166.89 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: F₃Fe·3H₂O
MW: 166.89 g/mol
MDL Number: MFCD00149718
CAS Number: 15469-38-2
EINECS: 232-002-4
UN: 3260
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR