Cyanoacetic acid ≥99%

Supplier: BeanTown Chemical

213025-1KG 213025-250G 213025-5KG
BT213025-1KGEA 80 USD
BT213025-1KG BT213025-250G BT213025-5KG
Cyanoacetic acid ≥99%
Cyanoacetic acid

CAS: 372-09-8; EC No: 206-743-9; MDL No: MFCD00002677; RTECS: AG3675000 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: NCCH2CO2H; Molecular Formula: C3H3NO2; MW: 85.06 Melting Point: 64-70°; Boiling Point: 108°/0.15 mmHg; Flash point: 107°C (225°F) Hygroscopic


  • RTECS: AG3675000
  • Hygroscopic
  • Crystalline

Formula: C₃H₃NO₂
MW: 85.06 g/mol
Boiling Pt: 108 °C/0.15 mmHg
Melting Pt: 64…70 °C
Flash Pt: 107 °C (225 °F)
MDL Number: MFCD00002677
CAS Number: 372-09-8
EINECS: 206-743-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR