2,6-Dibromopyridine ≥98%

Supplier: BeanTown Chemical

212990-100G 212990-25G 212990-500G
BT212990-100GEA 176 USD
BT212990-100G BT212990-25G BT212990-500G
2,6-Dibromopyridine ≥98%
2,6-Dibromopyridine

CAS: 626-05-1; EC No: 210-926-9; MDL No: MFCD00006223; RTECS: US7883000 UN No: UN2811; Haz Class: 6.1; Packing Group: II Crystalline; Molecular Formula: C5H3Br2N; MW: 236.89 Melting Point: 117-119°; Flash point: 100°C (212°F)


  • RTECS: US7883000
  • Crystalline

Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Melting Pt: 117…119 °C
Flash Pt: 100 °C (212 °F)
MDL Number: MFCD00006223
CAS Number: 626-05-1
EINECS: 210-926-9
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR