4-Iodobenzonitrile ≥98%
Supplier: BeanTown Chemical
Synonyms:
p-Iodobenzonitrile
212985-100G
212985-25G
212985-5G
BT212985-100GEA
1217
USD
BT212985-100G
BT212985-25G
BT212985-5G
4-Iodobenzonitrile ≥98%
4-Iodobenzonitrile
CAS: 3058-39-7; MDL No: MFCD00051310
UN No: UN3439; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C7H4IN ; MW: 229.02
Melting Point: 124-128°
Light Sensitive
- Light Sensitive
- Solid
Formula:
C₇H₄IN MW: 229.02 g/mol Melting Pt: 124…128 °C |
MDL Number:
MFCD00051310 CAS Number: 3058-39-7 UN: 3439 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...