Hafnium carbide ≥99.5%
Supplier: BeanTown Chemical
212980-10G
212980-50G
BT212980-10GEA
74.16
USD
BT212980-10G
BT212980-50G
Hafnium carbide ≥99.5%
Hafnium carbide
CAS: 12069-85-1; EC No: 235-114-1; MDL No: MFCD00016126; RTECS: MG4620000
UN No: UN3178; Haz Class: 4.1; Packing Group: III
-325 Mesh Powder; Linear Formula: HfC; MW: 190.50
Melting Point: 2930°
Density (g/mL): 12.2
- RTECS: MG4620000
- -325 Mesh Powder
Formula:
CHf MW: 190.54 g/mol Melting Pt: 2930 °C Density: 12.2 g/mL |
MDL Number:
MFCD00016126 CAS Number: 12069-85-1 EINECS: 235-114-1 UN: 3178 ADR: 4.1 III |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...