Nickel(II) formate dihydrate

Supplier: BeanTown Chemical

212970-100G 212970-2KG 212970-500G
BT212970-100GEA 32 USD
BT212970-100G BT212970-2KG BT212970-500G
Nickel(II) formate dihydrate
Nickel(II) formate dihydrate

CAS: 15694-70-9; EC No: 222-101-0; MDL No: MFCD00153034 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Molecular Formula: Ni(HCO2)2·2H2O; MW: 184.77 Density (g/mL): 2.15


  • Powder

Formula: Ni(HCO₂)₂·2H₂O
MW: 184.77 g/mol (148.74 anhy)
Density: 2.15 g/mL
MDL Number: MFCD00153034
CAS Number: 15694-70-9
EINECS: 222-101-0
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR