α-Bromo-2-iodotoulene ≥96%

Supplier: BeanTown Chemical

Synonyms: α-Bromo-o-iodotoulene, 2-Iodobenzyl bromide, 1-(Bromomethyl)-2-iodobenzene

212965-100G 212965-25G 212965-5G
BT212965-100GEA 965 USD
BT212965-100G BT212965-25G BT212965-5G
α-Bromo-2-iodotoulene ≥96%
α-Bromo-2-iodotoulene

CAS: 40400-13-3; MDL No: MFCD00236046 UN No: UN3261; Haz Class: 8; Packing Group: II Solid; Molecular Formula: C7H6BrI; MW: 296.93 Melting Point: 54-60° Light Sensitive


  • Light Sensitive
  • Solid

Formula: C₇H₆BrI
MW: 296.93 g/mol
Melting Pt: 54…60 °C
MDL Number: MFCD00236046
CAS Number: 40400-13-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR