Mercury(II) sulphate ≥98.0% ACS

Supplier: BeanTown Chemical

Synonyms: Mercury sulphate, Mercuric sulphate

212940-100G 212940-25G 212940-500G
BT212940-100GEA 114.81 USD
BT212940-100G BT212940-25G BT212940-500G
Mercury(II) sulphate ≥98.0% ACS
Mercury(II) sulphate

CAS: 7783-35-9; EC No: 231-992-5; MDL No: MFCD00011047; RTECS: OX0500000 UN No: UN1645; Haz Class: 6.1; Packing Group: II Powder; Linear Formula: HgSO4; MW: 296.65 Density (g/mL): 6.47 Air Sensitive, Light Sensitive, Moisture Sensitive


  • RTECS: OX0500000
  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • Powder

Formula: HgSO₄
MW: 296.65 g/mol
Melting Pt: 850
Density: 6.47 g/mL
MDL Number: MFCD00011047
CAS Number: 7783-35-9
EINECS: 231-992-5
UN: 1645
ADR: 6.1 II
Merck Index: 13,05914

Order Now

Specification Test Results

Assay 98.0% min.
Residue after reduction ≤0.02%
Iron (Fe) ≤0.005%
Chloride (Cl) ≤0.003%
Nitrate (NO3) Passes test
Mercurous mercury (as Hg) ≤0.15%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR