Potassium tricyanomethanide

Supplier: BeanTown Chemical

212920-1G 212920-5G
BT212920-1GEA 177 USD
BT212920-1G BT212920-5G
Potassium tricyanomethanide
Potassium tricyanomethanide

CAS: 34171-69-2; MDL No: MFCD00058850 UN No: UN3439; Haz Class: 6.1; Packing Group: II Crystalline; Linear Formula: KC(CN)3; MW: 129.16


  • Crystalline

Formula: C₄KN₃
MDL Number: MFCD00058850
CAS Number: 34171-69-2
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR