Zinc trifluoroacetate hydrate
Supplier: BeanTown Chemical
212830-100G
212830-25G
BT212830-100GEA
207
USD
BT212830-100G
BT212830-25G
Zinc trifluoroacetate hydrate
Zinc trifluoroacetate hydrate
CAS: 207801-31-8; EC No: 258-922-6; MDL No: MFCD00061595
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline/Powder; Linear Formula: Zn(CF3COO)2·xH2O; MW: 291.42 (anhydrous)
Hygroscopic
- Hygroscopic
- Crystalline/Powder
Formula:
C₄H₂F₆O₅Zn |
MDL Number:
MFCD00061595 CAS Number: 207801-31-8 EINECS: 258-922-6 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...