(S)-(-)-4-Methoxy-α-methylbenzylamine ≥99%, ee 99% ee

Supplier: BeanTown Chemical

Synonyms: (S)-1-(4-Methoxyphenyl)ethylamine, (S)-(-)-1-(4-Methoxyphenyl)ethylamine

212765-1G 212765-25G 212765-5G
BT212765-1GEA 35 USD
BT212765-1G BT212765-25G BT212765-5G
(S)-(-)-4-Methoxy-α-methylbenzylamine ≥99%, ee 99% ee
(S)-(-)-4-Methoxy-α-methylbenzylamine

CAS: 41851-59-6; MDL No: MFCD00671660 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C9H13NO; MW: 151.21 Density (g/mL): 1.024; Refractive Index: 1.533; Optical Rotation: [α]20/D +32±2°, neat Air Sensitive


  • Optical rotation: [α]20/D +32±2°, neat
  • Refractive index: 1.533
  • Air Sensitive
  • Liquid

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Density: 1.024 g/mL
MDL Number: MFCD00671660
CAS Number: 41851-59-6
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR