Ferrocene ≥99%

Supplier: BeanTown Chemical

212690-1KG 212690-250G 212690-50G
BT212690-1KGEA 223 USD
BT212690-1KG BT212690-250G BT212690-50G
Ferrocene ≥99%
Ferrocene

CAS: 102-54-5; EC No: 203-039-3; MDL No: MFCD00001427; RTECS: LK0700000 UN No: UN1325; Haz Class: 4.1; Packing Group: II Powder; Linear Formula: Fe(C5H5)2; Molecular Formula: C10H10Fe ; MW: 186.03 Melting Point: 172-174°; Boiling Point: 249° Air Sensitive, Moisture Sensitive


  • RTECS: LK0700000
  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: C₁₀H₁₀Fe
MW: 186.04 g/mol
Boiling Pt: 249 °C
Melting Pt: 172…174 °C
MDL Number: MFCD00001427
CAS Number: 102-54-5
EINECS: 203-039-3
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR