Terephthaloyl dichloride ≥99%
Supplier: BeanTown Chemical
Synonyms:
Terephthaloyl chloride
212605-1KG
212605-250G
BT212605-1KGEA
114
USD
BT212605-1KG
BT212605-250G
Terephthaloyl dichloride ≥99%
Terephthaloyl dichloride
CAS: 100-20-9; EC No: 202-829-5; MDL No: MFCD00000693; RTECS: WZ1797000
UN No: UN2928; Haz Class: 6.1 (8); Packing Group: II
Flakes; Linear Formula: C6H4(COCl)2; Molecular Formula: C8H5Cl2O2; MW: 203.02
Melting Point: 79-81°; Boiling Point: 266°; Flash point: 180°C (356°F)
Moisture Sensitive
- RTECS: WZ1797000
- Moisture Sensitive
- Flakes
Formula:
C₈H₄Cl₂O₂ MW: 203.02 g/mol Boiling Pt: 266 °C Melting Pt: 79…81 °C Flash Pt: 180 °C (356 °F) |
MDL Number:
MFCD00000693 CAS Number: 100-20-9 EINECS: 202-829-5 UN: 2928 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...