2,2,4-Trimethylpentane ≥99.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
212580-1L
212580-4L
212580-500ML
BT212580-1LEA
60.27
USD
BT212580-1L
BT212580-4L
BT212580-500ML
2,2,4-Trimethylpentane ≥99.0% ACS
2,2,4-Trimethylpentane
CAS: 540-84-1; EC No: 208-759-1; MDL No: MFCD00008943; RTECS: SA3320000
UN No: UN1262; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)2CHCH2C(CH3)3; Molecular Formula: C8H18; MW: 114.23
Melting Point: -107°; Boiling Point: 98-99°; Flash point: -12°C (10°F)
Density (g/mL): 0.692; Refractive Index: 1.391
- RTECS: SA3320000
- Refractive index: 1.391
- Liquid
Formula:
(CH₃)₂CHCH₂C(CH₃)₃ MW: 114.23 g/mol Boiling Pt: 98…99 °C Melting Pt: –107 °C Density: 0.692 g/mL Flash Pt: –12 °C (10 °F) |
MDL Number:
MFCD00008943 CAS Number: 540-84-1 EINECS: 208-759-1 UN: 1262 ADR: 3,II |
Specification Test Results
Assay | 99.0% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...