3-(Trifluoromethyl)phenyltrimethylammonium hydroxide 5% (w/v) in methanol

Supplier: BeanTown Chemical

212570-25ML 212570-5ML
BT212570-25MLEA 311 USD
BT212570-25ML BT212570-5ML
3-(Trifluoromethyl)phenyltrimethylammonium hydroxide 5% (w/v) in methanol
3-(Trifluoromethyl)phenyltrimethylammonium hydroxide

CAS: 68254-41-1; EC No: 200-659-6; MDL No: MFCD00059473 UN No: UN2924; Haz Class: 3(8); Packing Group: II Liquid; Molecular Formula: C10H14F3NO; MW: 221.22


  • Liquid

Formula: C₁₀H₁₄F₃NO
MDL Number: MFCD00059473
CAS Number: 68254-41-1
EINECS: 200-659-6
ADR: 3(8),II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR