2-Cyclopenten-1-one ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Cyclopentenone, Cyclopenten-2-one, Cyclopent-2-enone
212505-25G
212505-5G
BT212505-25GEA
255
USD
BT212505-25G
BT212505-5G
2-Cyclopenten-1-one ≥98%
2-Cyclopenten-1-one
CAS: 930-30-3; EC No: 213-213-0; MDL No: MFCD00001401
UN No: UN1224; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C5H6O ; MW: 82.10
Boiling Point: 64-65°/19 mmHg; Flash point: 42°C (108°F)
Density (g/mL): 0.98; Refractive Index: 1.481
- Refractive index: 1.481
- Liquid
Formula:
C₅H₆O MW: 82.1 g/mol Boiling Pt: 64…65 °C/19 mmHg Density: 0.98 g/mL Flash Pt: 42 °C (108 °F) |
MDL Number:
MFCD00001401 CAS Number: 930-30-3 EINECS: 213-213-0 UN: 1224 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...