Ethanesulfonyl chloride ≥98%
Supplier: BeanTown Chemical
CAS: 594-44-5; EC No: 209-842-5; MDL No: MFCD00007460; RTECS: KI8050000
UN No: UN2927; Haz Class: 6.1 (8); Packing Group: II
Liquid; Linear Formula: CH3CH2SO2Cl; Molecular Formula: C2H5ClO2S ; MW: 128.58
Boiling Point: 177°; Flash point: 83°C (181°F)
Density (g/mL): 1.357; Refractive Index: 1.452
Moisture Sensitive
- RTECS: KI8050000
- Refractive index: 1.452
- Moisture Sensitive
- Liquid
Formula:
C₂H₅ClO₂S Boiling Pt: 177 °C Density: 1.357 g/mL Flash Pt: 83 °C (181 °F) |
MDL Number:
MFCD00007460 CAS Number: 594-44-5 EINECS: 209-842-5 UN: 2927 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...