Sodium hexafluoroaluminate ≥97%

Supplier: BeanTown Chemical

Synonyms: Trisodium hexafluoroaluminate, Sodium hexafluoroaluminate(III)

212485-100G 212485-500G
BT212485-100GEA 30 USD
BT212485-100G BT212485-500G
Sodium hexafluoroaluminate ≥97%
Sodium hexafluoroaluminate

CAS: 13775-53-6; EC No: 237-410-6; MDL No: MFCD00003507; RTECS: BD0075000 UN No: UN3260; Haz Class: 8; Packing Group: III Powder; Linear Formula: Na3AlF6; MW: 209.95 Melting Point: 1000° Density (g/mL): 2.90


  • RTECS: BD0075000
  • Powder

Formula: AlF₆Na₃
MW: 209.94 g/mol
Melting Pt: 1000 °C
Density: 2.90 g/mL
MDL Number: MFCD00003507
CAS Number: 13775-53-6
EINECS: 237-410-6
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR