Platinium(IV) oxide hydrate ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

212455-1G 212455-5G
BT212455-1GEA 162.72 USD
BT212455-1G BT212455-5G
Platinium(IV) oxide hydrate ≥99.9% (trace metals basis)
Platinium(IV) oxide hydrate

CAS: 52785-06-5; EC No: 215-223-0; MDL No: MFCD00066964; RTECS: TP2506020 UN No: UN1479; Haz Class: 5.1; Packing Group: II Powder; Linear Formula: PtO2·xH2O; MW: 227.09 (anhydrous) Melting Point: 450°


  • RTECS: TP2506020
  • Powder

Formula: O₂Pt·H₂O
Melting Pt: 450 °C
MDL Number: MFCD00066964
CAS Number: 52785-06-5
EINECS: 215-223-0
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR