Zinc chloride hydrate ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
212440-10G
212440-250G
212440-50G
BT212440-10GEA
45.44
USD
BT212440-10G
BT212440-250G
BT212440-50G
Zinc chloride hydrate ≥99.99% (trace metals basis)
Zinc chloride hydrate
CAS: 7646-85-7; EC No: 231-592-0; MDL No: MFCD00011295
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline/Lump; Linear Formula: ZnCl2·xH2O; MW: 136.29 (anhydrous)
- Crystalline/Lump
Formula:
Cl₂Zn·H₂O |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 ADR: 8 III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...