Dodecyltrimethylammonium bromide ≥99%

Supplier: BeanTown Chemical

Synonyms: Dodecyl trimethyl ammonium bromide, (1-Dodecyl)trimethylammonium bromide

212435-10G 212435-250G 212435-50G
BT212435-10GEA 53.3 USD
BT212435-10G BT212435-250G BT212435-50G
Dodecyltrimethylammonium bromide ≥99%
Dodecyltrimethylammonium bromide

CAS: 1119-94-4; EC No: 214-290-3; MDL No: MFCD00011767; RTECS: BQ3195000 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline; Linear Formula: CH3(CH2)11N(Br)(CH3)3; Molecular Formula: C15H34BrN; MW: 308.34 Melting Point: 246° (decomposes) Hygroscopic


  • RTECS: BQ3195000
  • Hygroscopic
  • Crystalline

Formula: C₁₅H₃₄BrN
MW: 308.35 g/mol
Melting Pt: 246 °C (dec.)
MDL Number: MFCD00011767
CAS Number: 1119-94-4
EINECS: 214-290-3
ADR: 9,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR