1-Aminocyclopentanecarboxylic acid ≥97%

Supplier: BeanTown Chemical

212410-1G 212410-25G 212410-5G
BT212410-1GEA 38 USD
BT212410-1G BT212410-25G BT212410-5G
1-Aminocyclopentanecarboxylic acid ≥97%
1-Aminocyclopentanecarboxylic acid

CAS: 52-52-8; MDL No: MFCD00001381; RTECS: GY2625000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C6H11NO2 ; MW: 129.16 Melting Point: 320° (decomposes)


  • RTECS: GY2625000
  • Crystalline

Formula: C₆H₁₁NO₂
Melting Pt: 320 °C (dec.)
MDL Number: MFCD00001381
CAS Number: 52-52-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR