Dess-Martin periodinane

Supplier: BeanTown Chemical

Synonyms: 1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one, DMP, (1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl ester

212395-1G 212395-5G
BT212395-1GEA 37 USD
BT212395-1G BT212395-5G
Dess-Martin periodinane
Dess-Martin periodinane

CAS: 87413-09-0; MDL No: MFCD00130127 UN No: UN1479; Haz Class: 5.1; Packing Group: III Crystalline/Powder; Molecular Formula: C13H13IO8; MW: 424.15 Melting Point: 130-133° Light Sensitive, Moisture Sensitive


  • Light Sensitive, Moisture Sensitive
  • Crystalline/Powder

Formula: C₁₃H₁₃IO₈
MW: 424.15 g/mol
Melting Pt: 130…133 °C
MDL Number: MFCD00130127
CAS Number: 87413-09-0
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR