Benzyl benzoate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Benzoic acid benzyl ester
212375-1KG
212375-250G
BT212375-1KGEA
43
USD
BT212375-1KG
BT212375-250G
Benzyl benzoate ≥99%
Benzyl benzoate
CAS: 120-51-4; EC No: 204-402-9; MDL No: MFCD00003075; RTECS: DG4200000
UN No: UN3082; Haz Class: 9; Packing Group: III
Liquid; Molecular Formula: C14H12O2; MW: 212.24
Melting Point: 17-20°; Boiling Point: 323-324°; Flash point: 158°C (316°F)
Density (g/mL): 1.118; Refractive Index: 1.568
- RTECS: DG4200000
- Refractive index: 1.568
- Liquid
Formula:
C₁₄H₁₂O₂ MW: 212.25 g/mol Boiling Pt: 323…324 °C Melting Pt: 17…20 °C Density: 1.118 g/mL Flash Pt: 158 °C (316 °F) |
MDL Number:
MFCD00003075 CAS Number: 120-51-4 EINECS: 204-402-9 UN: 3082 ADR: 9,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...