2,6-Difluorobenzonitrile ≥98%
Supplier: BeanTown Chemical
212370-10G
212370-250G
212370-50G
BT212370-10GEA
27
USD
BT212370-10G
BT212370-250G
BT212370-50G
2,6-Difluorobenzonitrile ≥98%
2,6-Difluorobenzonitrile
CAS: 1897-52-5; EC No: 217-589-7; MDL No: MFCD00001778
UN No: UN3439; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C7H3F2N; MW: 139.10
Melting Point: 25-28°; Boiling Point: 197-198°; Flash point: 90°C (194°F)
Density (g/mL): 1.246; Refractive Index: 1.488
- Refractive index: 1.488
- Solid
Formula:
C₇H₃F₂N MW: 139.1 g/mol Boiling Pt: 197…198 °C Melting Pt: 25…28 °C Density: 1.246 g/mL Flash Pt: 90 °C (194 °F) |
MDL Number:
MFCD00001778 CAS Number: 1897-52-5 EINECS: 217-589-7 UN: 3439 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...