α,α,α-Trifluoro-3,4-diaminotoluene ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-(Trifluoromethyl)-1,2-phenylenediamine, 4-(Trifluoromethyl)benzene-1,2-diamine, 3,4-Diaminobenzotrifluoride, 4-(Trifluoromethyl)-o-phenylenediamine
212325-1G
212325-25G
212325-5G
BT212325-1GEA
40
USD
BT212325-1G
BT212325-25G
BT212325-5G
α,α,α-Trifluoro-3,4-diaminotoluene ≥98%
α,α,α-Trifluoro-3,4-diaminotoluene
CAS: 368-71-8; MDL No: MFCD00042456
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C7H7F3N2; MW: 176.14
Melting Point: 58-62°
- Powder
Formula:
C₇H₇F₃N₂ Melting Pt: 58…62 °C |
MDL Number:
MFCD00042456 CAS Number: 368-71-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...