N-(2-Aminoethyl)morpholine ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-(2-Aminoethyl)morpholine, 2-Morpholinoethanamine

212295-1KG 212295-250G 212295-50G
BT212295-1KGEA 589 USD
BT212295-1KG BT212295-250G BT212295-50G
N-(2-Aminoethyl)morpholine ≥98%
N-(2-Aminoethyl)morpholine

CAS: 2038-03-1; EC No: 218-011-6; MDL No: MFCD00006182; RTECS: QD7350000 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: II Solid; Molecular Formula: C6H14N2O; MW: 130.19 Boiling Point: 205°; Flash point: 86°C (187°F) Density (g/mL): 0.992; Refractive Index: 1.476 Air Sensitive


  • RTECS: QD7350000
  • Refractive index: 1.476
  • Air Sensitive
  • Solid

Formula: C₆H₁₄N₂O
MW: 130.19 g/mol
Boiling Pt: 205 °C
Density: 0.992 g/mL
Flash Pt: 86 °C (187 °F)
MDL Number: MFCD00006182
CAS Number: 2038-03-1
EINECS: 218-011-6
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR