(±)-3-Buten-2-ol ≥97%

Supplier: BeanTown Chemical

Synonyms: (±)-Methyl vinyl carbinol, (±)-3-Hydroxy-1-butene, 1-Buten-3-ol

212270-25G 212270-5G
BT212270-25GEA 88 USD
BT212270-25G BT212270-5G
(±)-3-Buten-2-ol ≥97%
(±)-3-Buten-2-ol

CAS: 598-32-3; EC No: 209-929-8; MDL No: MFCD00004543; RTECS: EM9275000 UN No: UN1986; Haz Class: 3 (6.1); Packing Group: II Liquid; Molecular Formula: C4H8O; MW: 72.11 Boiling Point: 96-97°; Flash point: 16°C (61°F) Density (g/mL): 0.832; Refractive Index: 1.415


  • RTECS: EM9275000
  • Refractive index: 1.415
  • Liquid

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 96…97 °C
Density: 0.832 g/mL
Flash Pt: 16 °C (61 °F)
MDL Number: MFCD00004543
CAS Number: 598-32-3
EINECS: 209-929-8
UN: 1986
ADR: 3,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR