Beryllium oxide ≥99% (trace metals basis)

Supplier: BeanTown Chemical

212175-25G 212175-5G
BT212175-25GEA 165 USD
BT212175-25G BT212175-5G
Beryllium oxide ≥99% (trace metals basis)
Beryllium oxide

CAS: 1304-56-9; EC No: 215-133-1; MDL No: MFCD00003457 UN No: UN1566; Haz Class: 6.1; Packing Group: II Powder; Molecular Formula: BeO; MW: 25.01 Density (g/mL): 3.01


  • Powder

Formula: BeO
Density: 3.01 g/mL
MDL Number: MFCD00003457
CAS Number: 1304-56-9
EINECS: 215-133-1
UN: 1566
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR