3-Chloropyridine ≥99%

Supplier: BeanTown Chemical

212105-10G 212105-50G
BT212105-10GEA 30 USD
BT212105-10G BT212105-50G
3-Chloropyridine ≥99%
3-Chloropyridine

CAS: 626-60-8; EC No: 210-955-7; MDL No: MFCD00006375; RTECS: US6125000 UN No: UN2924; Haz Class: 3(8); Packing Group: III Liquid; Molecular Formula: C5H4ClN ; MW: 113.54 Boiling Point: 148°; Flash point: 66°C (151°F) Density (g/mL): 1.194; Refractive Index: 1.533


  • RTECS: US6125000
  • Refractive index: 1.533
  • Liquid

Formula: C₅H₄ClN
MW: 113.55 g/mol
Boiling Pt: 148 °C
Density: 1.194 g/mL
Flash Pt: 66 °C (151 °F)
MDL Number: MFCD00006375
CAS Number: 626-60-8
EINECS: 210-955-7
UN: 2924
ADR: 3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR