Zinc phosphide ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Trizinc diphosphide, Zinc phosphide
212075-10G
212075-2G
212075-500MG
BT212075-10GEA
968
USD
BT212075-10G
BT212075-2G
BT212075-500MG
Zinc phosphide ≥99.9% (trace metals basis)
Zinc phosphide
CAS: 1314-84-7; EC No: 215-244-5; MDL No: MFCD00049631; RTECS: ZH4900000
UN No: UN1714; Haz Class: 4.3 (6.1); Packing Group: I
3mm & Down Agglomerated Powder; Linear Formula: Zn3P2; MW: 258.12
Melting Point: 420°; Boiling Point: 1100°
Density (g/mL): 4.550
Moisture Sensitive
- RTECS: ZH4900000
- Moisture Sensitive
- 3mm & Down Agglomerated Powder
Formula:
P₂Zn₃ MW: 258.12 g/mol Boiling Pt: 1100 °C Melting Pt: 420 °C Density: 4.550 g/mL |
MDL Number:
MFCD00049631 CAS Number: 1314-84-7 EINECS: 215-244-5 UN: 1714 ADR: 4.3,I |
Specification Test Results
Assay | 99.9% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...