p-Toluenesulphonyl isocyanate ≥95%
Supplier: BeanTown Chemical
Synonyms:
Tosyl isocyanate, 4-Isocyanatosulphonyltoluene
212065-100G
212065-25G
212065-500G
BT212065-100GEA
32
USD
BT212065-100G
BT212065-25G
BT212065-500G
p-Toluenesulphonyl isocyanate ≥95%
p-Toluenesulphonyl isocyanate
CAS: 4083-64-1; EC No: 223-810-8; MDL No: MFCD00002030; RTECS: DB9032000
UN No: UN2206; Haz Class: 6.1; Packing Group: III
Liquid; Linear Formula: CH3C6H4SO2NCO; Molecular Formula: C87NO3S; MW: 197.21
Melting Point: 5°; Boiling Point: 275-278°; Flash point: 145°C (293°F)
Density (g/mL): 1.291; Refractive Index: 1.534
Moisture Sensitive
- RTECS: DB9032000
- Refractive index: 1.534
- Moisture Sensitive
- Liquid
Formula:
C₈H₇NO₃S Boiling Pt: 275…278 °C Melting Pt: 5 °C Density: 1.291 g/mL Flash Pt: 145 °C (293 °F) |
MDL Number:
MFCD00002030 CAS Number: 4083-64-1 EINECS: 223-810-8 ADR: 6.1,III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...