Propanephosphonic acid cyclic anhydride 50% (w/w) in DMF

Supplier: BeanTown Chemical

Synonyms: 1-Propylphosphonic anhydride, 2,4,6-Tripropyl-1,3,5,2,4,6-trioxotriphosphorinane-2,4,6-trioxide

212040-10G 212040-50G
BT212040-10GEA 50.94 USD
BT212040-10G BT212040-50G
Propanephosphonic acid cyclic anhydride 50% (w/w) in DMF
Propanephosphonic acid cyclic anhydride

CAS: 68957-94-8; EC No: 422-210-5; MDL No: MFCD00006583 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C9H21O6P3; MW: 318.18 Flash point: 62°C (144°F) Density (g/mL): 1.09 Moisture Sensitive


  • Moisture Sensitive
  • Liquid

Formula: C₉H₂₁O₆P₃
MW: 318.19
Density: 1.09 g/mL
Flash Pt: 62 °C (144 °F)
MDL Number: MFCD00006583
CAS Number: 68957-94-8
EINECS: 422-210-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR