N,O-Bis(trimethylsilyl)acetamide ≥95%
Supplier: BeanTown Chemical
Synonyms:
BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate
212015-100G
212015-25G
212015-5G
BT212015-100GEA
116.29
USD
BT212015-100G
BT212015-25G
BT212015-5G
N,O-Bis(trimethylsilyl)acetamide ≥95%
N,O-Bis(trimethylsilyl)acetamide
CAS: 10416-59-8; EC No: 233-892-7; MDL No: MFCD00008270; RTECS: AK3000000
UN No: UN2920; Haz Class: 8 (3); Packing Group: II
Liquid/Low Melting Solid; Linear Formula: CH3C[=NSi(CH3)3]OSi(CH3)3; Molecular Formula: C8H21NOSi2; MW: 203.43
Melting Point: 24°; Boiling Point: 71-73°/35 mmHg; Flash point: 42°C (107°F)
Density (g/mL): 0.832; Refractive Index: 1.417
Air Sensitive, Moisture Sensitive
- RTECS: AK3000000
- Refractive index: 1.417
- Air Sensitive, Moisture Sensitive
- Liquid/Low Melting Solid
Formula:
C₈H₂₁NOSi₂ MW: 203.43 g/mol Boiling Pt: 71…73 °C/35 mmHg Melting Pt: 24 °C Density: 0.832 g/mL Flash Pt: 42 °C (107 °F) |
MDL Number:
MFCD00008270 CAS Number: 10416-59-8 EINECS: 233-892-7 UN: 2920 ADR: 8,II |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...