4,5,6,7-Tetrahydroisobenzofuran-1,3-dione ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-Cyclohexene-1,2-dicarboxylic anhydride, 3,4,5,6-Tetrahydrophthalic anhydride

211970-100G 211970-25G
BT211970-100GEA 351 USD
BT211970-100G BT211970-25G
4,5,6,7-Tetrahydroisobenzofuran-1,3-dione ≥97%
4,5,6,7-Tetrahydroisobenzofuran-1,3-dione

CAS: 2426-02-0; EC No: 219-374-3; MDL No: MFCD00005917 UN No: UN2698; Haz Class: 8; Packing Group: III Crystalline; Molecular Formula: C8H8O3; MW: 152.15 Melting Point: 69-73°


  • Crystalline

Formula: C₈H₈O₃
MW: 152.15
Melting Pt: 69…73 °C
MDL Number: MFCD00005917
CAS Number: 2426-02-0
EINECS: 219-374-3
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR