Methoxyacetic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Methoxyacetic acid

211960-1KG 211960-250G 211960-50G
BT211960-1KGEA 206 USD
BT211960-1KG BT211960-250G BT211960-50G
Methoxyacetic acid ≥97%
Methoxyacetic acid

CAS: 625-45-6; EC No: 210-894-6; MDL No: MFCD00004308; RTECS: AI8650000 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Linear Formula: CH3OCH2COOH; MW: 90.08 Melting Point: 7-9°; Boiling Point: 202-204°; Flash point: 121°C (250°F) Density (g/mL): 1.174; Refractive Index: 1.417


  • RTECS: AI8650000
  • Refractive index: 1.417
  • Liquid

Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 202…204 °C
Melting Pt: 7…9 °C
Density: 1.174 g/mL
Flash Pt: 121 °C (250 °F)
MDL Number: MFCD00004308
CAS Number: 625-45-6
EINECS: 210-894-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR