Perrhenic acid ≥78-80% (w/w) in water
Supplier: BeanTown Chemical
211945-25G
211945-5G
BT211945-25GEA
490
USD
BT211945-25G
BT211945-5G
Perrhenic acid ≥78-80% (w/w) in water
Perrhenic acid
CAS: 13768-11-1; EC No: 237-380-4; MDL No: MFCD00011326
UN No: UN3264; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: HReO4; MW: 251.21
- Liquid
Formula:
HO₄Re MW: 251.21 g/mol |
MDL Number:
MFCD00011326 CAS Number: 13768-11-1 EINECS: 237-380-4 UN: 3264 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...