4-Bromo-2,6-xylidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Bromo-2,6-dimethylaniline

211930-10G 211930-50G
BT211930-10GEA 34 USD
BT211930-10G BT211930-50G
4-Bromo-2,6-xylidine ≥98%
4-Bromo-2,6-xylidine

CAS: 24596-19-8; EC No: 246-337-9; MDL No: MFCD00007826 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C8H10BrN; MW: 200.08 Melting Point: 48-51°; Flash point: 110°C (230°F)


  • Crystalline

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Melting Pt: 48…51 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00007826
CAS Number: 24596-19-8
EINECS: 246-337-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR