Barium sulfide ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
211925-10G
211925-50G
BT211925-10GEA
86
USD
BT211925-10G
BT211925-50G
Barium sulfide ≥99.5% (trace metals basis)
Barium sulphide
CAS: 21109-95-5; EC No: 244-214-4; MDL No: MFCD00014192
UN No: UN2813; Haz Class: 4.3; Packing Group: II
-200 Mesh Powder ; Molecular Formula: BaS ; MW: 163.39
Density (g/mL): 4.25
Moisture Sensitive
- Moisture Sensitive
- -200 Mesh Powder
Formula:
BaS Density: 4.25 g/mL |
MDL Number:
MFCD00014192 CAS Number: 21109-95-5 EINECS: 244-214-4 UN: 2813 ADR: 4.3,II |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...