Digallium trisulfide ≥99.99% (trace metals basis) may contain small amounts of oxide

Supplier: BeanTown Chemical

Synonyms: Gallium (III) sulphide

211885-10G 211885-2G 211885-500MG
BT211885-10GEA 299 USD
BT211885-10G BT211885-2G BT211885-500MG
Digallium trisulfide ≥99.99% (trace metals basis) may contain small amounts of oxide
Digallium trisulphide

CAS: 12024-22-5; EC No: 234-692-2; MDL No: MFCD00016106 UN No: UN3260; Haz Class: 8; Packing Group: II Powder; Molecular Formula: Ga2S3; MW: 235.63 Density (g/mL): 3.65 Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: Ga₂S₃
MW: 235.64 g/mol
Density: 3.65 g/mL
MDL Number: MFCD00016106
CAS Number: 12024-22-5
EINECS: 234-692-2
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR