Zinc oxide 99,999

Supplier: BeanTown Chemical

Synonyms: Zinci oxidum, Zinc (II) oxide

211845-10G 211845-250G 211845-50G
BT211845-10GEA 50 USD
BT211845-10G BT211845-250G BT211845-50G
Zinc oxide 99,999
Zinc oxide

CAS: 1314-13-2; EC No: 215-222-5; MDL No: MFCD00011300; RTECS: ZH4810000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: ZnO; MW: 81.39 Melting Point: 1975° Density (g/mL): 5.61


  • RTECS: ZH4810000
  • Powder

Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.61 g/mL
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR