1-Octene ≥97%

Supplier: BeanTown Chemical

211780-2.5L 211780-500ML
BT211780-2.5LEA 95.34 USD
BT211780-2.5L BT211780-500ML
1-Octene ≥97%
1-Octene

CAS: 111-66-0; EC No: 203-893-7; MDL No: MFCD00009548; RTECS: RH2207000 UN No: UN3295; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3(CH2)5CH=CH2; Molecular Formula: C8H16; MW: 112.21 Melting Point: -101°; Boiling Point: 122-123°; Flash point: 10°C (50°F) Density (g/mL): 0.715; Refractive Index: 1.409


  • RTECS: RH2207000
  • Refractive index: 1.409
  • Liquid

Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 122…123 °C
Melting Pt: –101 °C
Density: 0.715 g/mL
Flash Pt: 10 °C (50 °F)
MDL Number: MFCD00009548
CAS Number: 111-66-0
EINECS: 203-893-7
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR